Thermo Scientific Chemicals

2-Chlorobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B23324.14
25 g, Each
Thermo Scientific Chemicals

2-Chlorobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B23324.14
25 g, Each
Quantity
Catalog number: B23324.14
also known as B23324-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3900-89-8
IUPAC Name
(2-chlorophenyl)boronic acid
Molecular Formula
C6H6BClO2
InChI Key
RRCMGJCFMJBHQC-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=CC=C1Cl
Assay (HPLC)
≥96.0%
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder

Description

2-Chlorobenzeneboronic acid is used in the preparation of imidazo[1,2-a]pyridine amides which has tuberculostatic activity. It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzeneboronic acid is used in the preparation of imidazo[1,2-a]pyridine amides which has tuberculostatic activity. It is used as pharmaceutical intermediates.

Solubility
Soluble in DMSO, Methanol.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text