2-Ethoxynaphthalene, 99%, Thermo Scientific Chemicals
2-Ethoxynaphthalene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Ethoxynaphthalene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number B23327.22
also known as B23327-22
Price (USD)/ Each
41.50
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
41.50
Add to cart
2-Ethoxynaphthalene, 99%, Thermo Scientific Chemicals
Catalog numberB23327.22
Price (USD)/ Each
41.50
-
Add to cart
Chemical Identifiers
CAS93-18-5
IUPAC Name2-ethoxynaphthalene
Molecular FormulaC12H12O
InChI KeyGUMOJENFFHZAFP-UHFFFAOYSA-N
SMILESCCOC1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)33.5-39.5?C
Appearance (Color)White
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only