2-Methyl-1,3-dithiolane, 99%, Thermo Scientific Chemicals
2-Methyl-1,3-dithiolane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyl-1,3-dithiolane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B23372.06
also known as B23372-06
Price (USD)/ Each
64.50
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
64.50
Add to cart
2-Methyl-1,3-dithiolane, 99%, Thermo Scientific Chemicals
Catalog numberB23372.06
Price (USD)/ Each
64.50
-
Add to cart
Chemical Identifiers
CAS5616-51-3
IUPAC Name2-methyl-1,3-dithiolane
Molecular FormulaC4H8S2
InChI KeyCARJCVDELAMAEJ-UHFFFAOYSA-N
SMILESCC1SCCS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
FormLiquid
Refractive Index1.5625-1.5655 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only