2-Amino-6-methoxybenzothiazole, 98%, Thermo Scientific Chemicals
2-Amino-6-methoxybenzothiazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-6-methoxybenzothiazole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number B23380.18
also known as B23380-18
Price (USD)/ Each
98.60
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
98.60
Add to cart
2-Amino-6-methoxybenzothiazole, 98%, Thermo Scientific Chemicals
Catalog numberB23380.18
Price (USD)/ Each
98.60
-
Add to cart
Chemical Identifiers
CAS1747-60-0
IUPAC Name6-methoxy-1,3-benzothiazol-2-amine
Molecular FormulaC8H8N2OS
InChI KeyKZHGPDSVHSDCMX-UHFFFAOYSA-N
SMILESCOC1=CC=C2N=C(N)SC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pale brown
Assay (GC)≥97.5%
Melting Point (clear melt)158.0-165.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only