Thermo Scientific Chemicals

4-Chloro-2-nitrotoluene, 99%, Thermo Scientific Chemicals

Catalog number: B23382.36
500 g, Each
Thermo Scientific Chemicals

4-Chloro-2-nitrotoluene, 99%, Thermo Scientific Chemicals

Catalog number: B23382.36
500 g, Each
Quantity
Catalog number: B23382.36
also known as B23382-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
89-59-8
IUPAC Name
4-chloro-1-methyl-2-nitrobenzene
Molecular Formula
C7H6ClNO2
InChI Key
SQFLFRQWPBEDHM-UHFFFAOYSA-N
SMILES
CC1=CC=C(Cl)C=C1[N+]([O-])=O
Melting Point (clear melt)
34.0-40.0?C
Appearance (Color)
Pale yellow to yellow to pale brown
Assay (GC)
≥98.5%
Form
Crystals or powder or crystalline powder or lumps or fused solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text