Triethyl 1,3,5-benzenetricarboxylate, 97%, Thermo Scientific Chemicals
Triethyl 1,3,5-benzenetricarboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethyl 1,3,5-benzenetricarboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B23392.14
also known as B23392-14
Price (USD)/ Each
164.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
164.00
Add to cart
Triethyl 1,3,5-benzenetricarboxylate, 97%, Thermo Scientific Chemicals
Catalog numberB23392.14
Price (USD)/ Each
164.00
-
Add to cart
Chemical Identifiers
CAS4105-92-4
IUPAC Name1,3,5-triethyl benzene-1,3,5-tricarboxylate
Molecular FormulaC15H18O6
InChI KeyKXGOWZRHSOJOLF-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC(=CC(=C1)C(=O)OCC)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)129.5-138.5?C
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only