Thermo Scientific Chemicals

1,2-Benzisoxazole, 95%, Thermo Scientific Chemicals

Catalog number: B23400.06
5 g, Each
Thermo Scientific Chemicals

1,2-Benzisoxazole, 95%, Thermo Scientific Chemicals

Catalog number: B23400.06
5 g, Each
Quantity
Catalog number: B23400.06
also known as B23400-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
271-95-4
IUPAC Name
1,2-benzoxazole
Molecular Formula
C7H5NO
InChI Key
KTZQTRPPVKQPFO-UHFFFAOYSA-N
SMILES
O1N=CC2=CC=CC=C12
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥94.0%
Refractive Index
1.5590-1.5650 @ 20?C

Description

1,2-Benzisoxazole derivative zonisamide is a novel antiepileptic drug and is effective for the treatment of partial seizures. 1,2-Benzisoxazole is a potential substrates of rabbit liver aldehyde oxidase. It is used to produce 5-nitro-benzo[d]isoxazole. It is involved in manufacturing medicines ( antiepileptics and antipsychotics).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water. 4.1 mg/mL in water at 25°C

Notes
Light Sensitive. Store at 4°C. Store in the dark. Incompatible with light and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text