Thermo Scientific Chemicals

3-Amino-2,4,6-triiodobenzoic acid, 99% (dry wt.), water <4%, Thermo Scientific Chemicals

Catalog number: B23405.06
5 g, Each
Thermo Scientific Chemicals

3-Amino-2,4,6-triiodobenzoic acid, 99% (dry wt.), water <4%, Thermo Scientific Chemicals

Catalog number: B23405.06
5 g, Each
Quantity
Catalog number: B23405.06
also known as B23405-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3119-15-1
IUPAC Name
sodium 3-amino-2,4,6-triiodobenzoate
Molecular Formula
C7H3I3NNaO2
InChI Key
CTXRMWPLTDAHOR-UHFFFAOYSA-M
SMILES
[Na+].NC1=C(I)C=C(I)C(C([O-])=O)=C1I
Appearance (Color)
White to pale cream to pale brown
Form
Powder
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5% (dry wt. basis)
Assay (HPLC)
≥98.5% (dry wt. basis)
Identification (FTIR)
Conforms

Description

3-Amino-2,4,6-triiodobenzoic acid is used to prepare polymeric hydrogel based particles used in vascular embolization. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-2,4,6-triiodobenzoic acid is used to prepare polymeric hydrogel based particles used in vascular embolization. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Protect from exposure to light. Store in the dark. Incompatible with light and oxidizing agents. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text