Thermo Scientific Chemicals

2,2'-Dimethoxybiphenyl, 97%, Thermo Scientific Chemicals

Catalog number: B23411.22
100 g, Each
Thermo Scientific Chemicals

2,2'-Dimethoxybiphenyl, 97%, Thermo Scientific Chemicals

Catalog number: B23411.22
100 g, Each
Quantity
Catalog number: B23411.22
also known as B23411-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4877-93-4
IUPAC Name
2,2'-dimethoxy-1,1'-biphenyl
Molecular Formula
C14H14O2
InChI Key
VGMKUVCDINAAFC-UHFFFAOYSA-N
SMILES
COC1=CC=CC=C1C1=CC=CC=C1OC
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
151.0-160.0?C
Assay (GC)
≥96.0%
Appearance (Color)
White to pale cream

Description

2,2-Dimethoxy-1,1-biphenyl is used in the synthesis of biphenyl scaffolds in the preparation of biphenyl-tetrathiafulvalene derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dimethoxy-1,1-biphenyl is used in the synthesis of biphenyl scaffolds in the preparation of biphenyl-tetrathiafulvalene derivatives.

Solubility
Soluble in chloroform, methanol.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text