Thermo Scientific Chemicals

1,8-Dichloroanthraquinone, 96%, Thermo Scientific Chemicals

Catalog number: B23421.14
25 g, Each
Thermo Scientific Chemicals

1,8-Dichloroanthraquinone, 96%, Thermo Scientific Chemicals

Catalog number: B23421.14
25 g, Each
Quantity
Catalog number: B23421.14
also known as B23421-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
82-43-9
IUPAC Name
1,8-dichloro-9,10-dihydroanthracene-9,10-dione
Molecular Formula
C14H6Cl2O2
InChI Key
VBQNYYXVDQUKIU-UHFFFAOYSA-N
SMILES
ClC1=CC=CC2=C1C(=O)C1=C(Cl)C=CC=C1C2=O
Appearance (Color)
Pale yellow to yellow
Assay (GC)
≥95.0%
Form
Powder
Melting Point (clear melt)
199.0-208.0?C

Description

1,8-Dichloroanthraquinone is a reagent in the preparation of anthraquinones, which has antitumor activities. It is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Dichloroanthraquinone is a reagent in the preparation of anthraquinones, which has antitumor activities. It is used as primary and secondary intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text