Thermo Scientific Chemicals

3,5-Dimethylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: B23434.03
1 g, Each
Thermo Scientific Chemicals

3,5-Dimethylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: B23434.03
1 g, Each
Quantity
Catalog number: B23434.03
also known as B23434-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
172975-69-8
IUPAC Name
(3,5-dimethylphenyl)boronic acid
Molecular Formula
C8H11BO2
InChI Key
DJGHSJBYKIQHIK-UHFFFAOYSA-N
SMILES
CC1=CC(=CC(C)=C1)B(O)O
Form
Powder
Assay (Aqueous acid-base Titration)
≥97.5%
Assay (HPLC)
≥97.5%
Proton NMR
Conforms to structure
Appearance (Color)
White

Description

3,5-Dimethylbenzeneboronic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylbenzeneboronic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text