Thermo Scientific Chemicals

Aminomethanesulfonic acid, 97%, Thermo Scientific Chemicals

Catalog number: B23441.36
500 g, Each
Thermo Scientific Chemicals

Aminomethanesulfonic acid, 97%, Thermo Scientific Chemicals

Catalog number: B23441.36
500 g, Each
Quantity
Catalog number: B23441.36
also known as B23441-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13881-91-9
IUPAC Name
aminomethanesulfonic acid
Molecular Formula
CH5NO3S
InChI Key
OBESRABRARNZJB-UHFFFAOYSA-N
SMILES
NCS(O)(=O)=O
Appearance (Color)
White
Form
powder
Assay (Aqueous acid-base Titration)
>96.0%

Description

Aminomethanesulfonic acid is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aminomethanesulfonic acid is used as a pharmaceutical intermediates.

Solubility
Soluble in water. (17 g/L) at 17°C

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text