1-Bromo-2-ethylbenzene, 98%, Thermo Scientific Chemicals
1-Bromo-2-ethylbenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromo-2-ethylbenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number B23449.22
also known as B23449-22
Price (USD)/ Each
457.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
457.00
Add to cart
1-Bromo-2-ethylbenzene, 98%, Thermo Scientific Chemicals
Catalog numberB23449.22
Price (USD)/ Each
457.00
-
Add to cart
Chemical Identifiers
CAS1973-22-4
IUPAC Name1-bromo-2-ethylbenzene
Molecular FormulaC8H9Br
InChI KeyHVRUGFJYCAFAAN-UHFFFAOYSA-N
SMILESCCC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.5460-1.5500 @ 20?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only