Thermo Scientific Chemicals

1-Formylpiperidine, 99%, Thermo Scientific Chemicals

Catalog number: B23452.30
250 g, Each
Thermo Scientific Chemicals

1-Formylpiperidine, 99%, Thermo Scientific Chemicals

Catalog number: B23452.30
250 g, Each
Quantity
Catalog number: B23452.30
also known as B23452-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2591-86-8
IUPAC Name
piperidine-1-carbaldehyde
Molecular Formula
C6H11NO
InChI Key
FEWLNYSYJNLUOO-UHFFFAOYSA-N
SMILES
O=CN1CCCCC1
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Refractive Index
1.4830-1.4860 @ 20?C
Form
Liquid

Description

1-Formylpiperidine reacts with Grignard or alkyllithium reagents gives aldehydes in high yield. It can be used as a polar aprotic solvent, with better hydrocarbon solubility than other amide solvents such as dimethylformamide. In some formylation reaction of alkyllithium compounds it gives higher yields than the DMF.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Formylpiperidine reacts with Grignard or alkyllithium reagents gives aldehydes in high yield. It can be used as a polar aprotic solvent, with better hydrocarbon solubility than other amide solvents such as dimethylformamide. In some formylation reaction of alkyllithium compounds it gives higher yields than the DMF.

Solubility
Freely soluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text