Thermo Scientific Chemicals

4-(Methylthio)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B23454.03
1 g, Each
Thermo Scientific Chemicals

4-(Methylthio)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B23454.03
1 g, Each
Quantity
Catalog number: B23454.03
also known as B23454-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98546-51-1
IUPAC Name
[4-(methylsulfanyl)phenyl]boronic acid
Molecular Formula
C7H9BO2S
InChI Key
IVUHTLFKBDDICS-UHFFFAOYSA-N
SMILES
CSC1=CC=C(C=C1)B(O)O
Appearance (Color)
White to pale yellow
Assay (HPLC)
≥96.0% (U.K. specification)
Comment
Purchased in U.K. and U.S.
Assay (Aqueous acid-base Titration)
≥96.0% (U.K. specification)
Assay from Supplier's CofA
≥96.0% (HPLC) (U.S. specification)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text