Chemical Identifiers
CAS2130-96-3
IUPAC Name(2S)-3-[4-(benzyloxy)phenyl]-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Molecular FormulaC21H25NO5
InChI KeyZAVSPTOJKOFMTA-LGWFVXIRNA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O
View more