Thermo Scientific Chemicals

Cobalt(II) thiocyanate, 98+%, Thermo Scientific Chemicals

Catalog number: B23460.30
250 g, Each
Thermo Scientific Chemicals

Cobalt(II) thiocyanate, 98+%, Thermo Scientific Chemicals

Catalog number: B23460.30
250 g, Each
Quantity
Catalog number: B23460.30
also known as B23460-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3017-60-5
IUPAC Name
λ²-cobalt(2+) bis(cyanosulfanide)
Molecular Formula
C2CoN2S2
InChI Key
INDBQWVYFLTCFF-UHFFFAOYSA-L
SMILES
[Co++].[S-]C#N.[S-]C#N
Appearance (Color)
Brown
Form
powder
Assay (unspecified)
>98.0%
Insoluble material
5% in water contains <1% insolubles

Description

Cobalt(II) thiocyanate is best known for Scott test, which is a proven screening test for the presence of cocaine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) thiocyanate is best known for Scott test, which is a proven screening test for the presence of cocaine.

Solubility
Soluble in water.

Notes
Incompatible with acids and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text