Thermo Scientific Chemicals

Benzyl acetate, 99%, Thermo Scientific Chemicals

Catalog number: B23472.36
500 g, Each
Thermo Scientific Chemicals

Benzyl acetate, 99%, Thermo Scientific Chemicals

Catalog number: B23472.36
500 g, Each
Quantity
Catalog number: B23472.36
also known as B23472-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
140-11-4
IUPAC Name
benzyl acetate
Molecular Formula
C9H10O2
InChI Key
QUKGYYKBILRGFE-UHFFFAOYSA-N
SMILES
CC(=O)OCC1=CC=CC=C1
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Refractive Index
1.5010-1.5040 @ 20?C

Description

Benzyl acetate can be used as a high boiling point solvent in coatings, like ink coating, binding agent and paint remover. It is used in soap and other chemical essences and has the effect of promote in floral and fantasy essences in jasmine, white orchid, fragrant plantain lily, gekkakou, narcissus and other essences. It is a plasticizer for ionophore membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl acetate can be used as a high boiling point solvent in coatings, like ink coating, binding agent and paint remover. It is used in soap and other chemical essences and has the effect of promote in floral and fantasy essences in jasmine, white orchid, fragrant plantain lily, gekkakou, narcissus and other essences. It is a plasticizer for ionophore membranes.

Solubility
Soluble in water 3.1 g/L.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text