2-Nitro-p-phenylenediamine, 95%, Thermo Scientific Chemicals
2-Nitro-p-phenylenediamine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Nitro-p-phenylenediamine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B23477.14
also known as B23477-14
Price (USD)/ Each
37.80
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
37.80
Add to cart
2-Nitro-p-phenylenediamine, 95%, Thermo Scientific Chemicals
Catalog numberB23477.14
Price (USD)/ Each
37.80
-
Add to cart
Chemical Identifiers
CAS5307-14-2
IUPAC Name2-nitrobenzene-1,4-diamine
Molecular FormulaC6H7N3O2
InChI KeyHVHNMNGARPCGGD-UHFFFAOYSA-N
SMILESNC1=CC=C(N)C(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Dark green to dark brown to black
Assay (Non-aqueous acid-base Titration)≥94.0 to ≤106.0%
Melting Point (clear melt)129.5-141.5?C
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only