Thermo Scientific Chemicals

3-Acetylbenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B23478.14
25 g, Each
Thermo Scientific Chemicals

3-Acetylbenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B23478.14
25 g, Each
Quantity
Catalog number: B23478.14
also known as B23478-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
204841-19-0
IUPAC Name
(3-acetylphenyl)boronic acid
Molecular Formula
C8H9BO3
InChI Key
SJGGDZCTGBKBCK-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=CC(=C1)B(O)O
Form
Powder
Assay (HPLC)
≥96.0%
Appearance (Color)
White to pale cream to pale yellow
Proton NMR
Conforms to structure
Assay (Aqueous acid-base Titration)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text