Cyclododecanol, 99%, Thermo Scientific Chemicals
Cyclododecanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclododecanol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Catalog number B23486.14
also known as B23486-14
Price (USD)/ Each
40.90
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
40.90
Add to cart
Cyclododecanol, 99%, Thermo Scientific Chemicals
Catalog numberB23486.14
Price (USD)/ Each
40.90
-
Add to cart
Chemical Identifiers
CAS1724-39-6
IUPAC Namecyclododecanol
Molecular FormulaC12H24O
InChI KeySFVWPXMPRCIVOK-UHFFFAOYSA-N
SMILESOC1CCCCCCCCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or fused/lumpy solid
Appearance (Color)White
Assay (GC)≥98.5%
Melting Point (clear melt)75.0-81.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only