Thermo Scientific Chemicals

Cyclododecanol, 99%, Thermo Scientific Chemicals

Catalog number: B23486.36
500 g, Each
Thermo Scientific Chemicals

Cyclododecanol, 99%, Thermo Scientific Chemicals

Catalog number: B23486.36
500 g, Each
Quantity
Catalog number: B23486.36
also known as B23486-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1724-39-6
IUPAC Name
cyclododecanol
Molecular Formula
C12H24O
InChI Key
SFVWPXMPRCIVOK-UHFFFAOYSA-N
SMILES
OC1CCCCCCCCCCC1
Form
Powder or fused/lumpy solid
Appearance (Color)
White
Assay (GC)
≥98.5%
Melting Point (clear melt)
75.0-81.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text