Thermo Scientific Chemicals

Benzyl methyl ether, 97%, Thermo Scientific Chemicals

Catalog number: B23490.09
10 g, Each
Thermo Scientific Chemicals

Benzyl methyl ether, 97%, Thermo Scientific Chemicals

Catalog number: B23490.09
10 g, Each
Quantity
Catalog number: B23490.09
also known as B23490-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
538-86-3
IUPAC Name
(methoxymethyl)benzene
Molecular Formula
C8H10O
InChI Key
GQKZBCPTCWJTAS-UHFFFAOYSA-N
SMILES
COCC1=CC=CC=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.4990-1.5040 @ 20?C
Identification (FTIR)
Conforms

Description

Benzyl methyl ether is used to produce benzaldehyde. It is used as perfuming agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl methyl ether is used to produce benzaldehyde. It is used as perfuming agents.

Solubility
Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text