Thermo Scientific Chemicals

n-Dodecylpyridinium chloride hydrate, 98%, Thermo Scientific Chemicals

Catalog number: B23492.22
100 g, Each
Thermo Scientific Chemicals

n-Dodecylpyridinium chloride hydrate, 98%, Thermo Scientific Chemicals

Catalog number: B23492.22
100 g, Each
Quantity
Catalog number: B23492.22
also known as B23492-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
207234-02-4
IUPAC Name
1-dodecylpyridin-1-ium chloride
Molecular Formula
C17H30ClN
InChI Key
GKQHIYSTBXDYNQ-UHFFFAOYSA-M
SMILES
[Cl-].CCCCCCCCCCCC[N+]1=CC=CC=C1
Assay (Titration ex Chloride)
≥97.5 to ≤102.5% (Dry wt. basis)
Form
Powder or crystalline powder
Melting Point (clear melt)
87-93?C
Water Content (Karl Fischer Titration)
<6.6% (<1.1 waters)
Appearance (Color)
White to pale cream

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text