Thermo Scientific Chemicals

2-Chloro-3',4'-dihydroxyacetophenone, 97%, Thermo Scientific Chemicals

Catalog number: B23496.14
25 g, Each
Thermo Scientific Chemicals

2-Chloro-3',4'-dihydroxyacetophenone, 97%, Thermo Scientific Chemicals

Catalog number: B23496.14
25 g, Each
Quantity
Catalog number: B23496.14
also known as B23496-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-40-1
IUPAC Name
2-chloro-1-(3,4-dihydroxyphenyl)ethan-1-one
Molecular Formula
C8H7ClO3
InChI Key
LWTJEJCZJFZKEL-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C1O)C(=O)CCl
Appearance (Color)
Grey or cream to brown or purple
Assay (HPLC)
>96.0%
Form
Powder

Description

2-Chloro-3',4'-dihydroxyacetophenone is used as a styptic adrenobazone, quasi adrenaline drug gasp spirit of intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-3′,4′-dihydroxyacetophenone is used as a styptic adrenobazone, quasi adrenaline drug gasp spirit of intermediates.

Solubility
Soluble in DMSO, methanol.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text