Thermo Scientific Chemicals

Dichloroisocyanuric acid sodium salt, 97% (dry wt.), Thermo Scientific Chemicals

Catalog number: B23504.36
500 g, Each
Thermo Scientific Chemicals

Dichloroisocyanuric acid sodium salt, 97% (dry wt.), Thermo Scientific Chemicals

Catalog number: B23504.36
500 g, Each
Quantity
Catalog number: B23504.36
also known as B23504-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2893-78-9
IUPAC Name
sodium 3,5-dichloro-2,4,6-trioxo-1,3,5-triazinan-1-ide
Molecular Formula
C3Cl2N3NaO3
InChI Key
MSFGZHUJTJBYFA-UHFFFAOYSA-M
SMILES
[Na+].ClN1C(=O)[N-]C(=O)N(Cl)C1=O
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0% (on dried substance)
Appearance (Color)
White
Form
Granules or granular powder or crystals
Identification (FTIR)
Conforms
Loss on Drying
≤5.0%

Description

Sodium dichloroisocyanurate can be used as a reagent for N-monochlorination and dehydrochlorination of amino esters. It is also a reagent for chlorination to detect ammonium via formation of colored zebra-bands in a detecting tube.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium dichloroisocyanurate can be used as a reagent for N-monochlorination and dehydrochlorination of amino esters, synthesis of antimicrobial polyurethane additives to control biofilm. It is used as a emergency disinfectant in drinking water, widely used cleansing agent. It is also a reagent for chlorination to detect ammonium via formation of colored zebra-bands in a detecting tube.

Solubility
Soluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text