Thermo Scientific Chemicals

4-(Aminomethyl)benzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: B23519.14
25 g, Each
Thermo Scientific Chemicals

4-(Aminomethyl)benzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: B23519.14
25 g, Each
Quantity
Catalog number: B23519.14
also known as B23519-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-91-7
IUPAC Name
4-(aminomethyl)benzoic acid
Molecular Formula
C8H9NO2
InChI Key
QCTBMLYLENLHLA-UHFFFAOYSA-N
SMILES
NCC1=CC=C(C=C1)C(O)=O
Appearance (Color)
White to cream
Assay (HPLC)
>96.0%
Water Content (Karl Fischer Titration)
≤4.0%
Form
Powder
Assay (Non-aqueous acid-base Titration)
>96.0% (dry wt. basis)

Description

4-(Aminomethyl)benzoic acid acts as an unnatural amino acid derivative. It is also used as a type 2 antifibrinolytic agent. Further, it reacts with 2-methyl-isothiourea sulfate to prepare 4-guanidinomethylbenzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Aminomethyl)benzoic acid acts as an unnatural amino acid derivative. It is also used as a type 2 antifibrinolytic agent. Further, it reacts with 2-methyl-isothiourea sulfate to prepare 4-guanidinomethylbenzoic acid.

Solubility
Slightly soluble in water. Insoluble in ethanol, benzene and chloroform.

Notes
Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text