Thermo Scientific Chemicals

(+)-Griseofulvin, 97%, Thermo Scientific Chemicals

Catalog number: B23521.22
100 g, Each
Thermo Scientific Chemicals

(+)-Griseofulvin, 97%, Thermo Scientific Chemicals

Catalog number: B23521.22
100 g, Each
Quantity
Catalog number: B23521.22
also known as B23521-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
126-07-8
IUPAC Name
(2S,2'R)-7-chloro-4,6,6'-trimethoxy-2'-methyl-3H-spiro[1-benzofuran-2,1'-cyclohexane]-3,4'-dione
Molecular Formula
C17H19ClO6
InChI Key
IIUZTXTZRGLYTI-LKUJNTHKSA-N
SMILES
COC1CC(=O)C[C@@H](C)[C@]11OC2=C(Cl)C(OC)=CC(OC)=C2C1=O
Appearance (Color)
White
Identification (FTIR)
Conforms (non-U.S. sourced material)
Proton NMR
Conforms to structure (U.S. sourced material)
Form
Powder
Comment
Material sourced in the U.S. and in other countries

Description

Griseofulvin is an antifungal drug. It is used both in animal and in humans, to treat ringworm infections of the skin and nails.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Griseofulvin is an antifungal drug. It is used both in animal and in humans, to treat ringworm infections of the skin and nails.

Solubility
Soluble in dimethyl formamide, ethanol, methanol, chloroform, DMSO, tetrachloroethane. Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text