Thermo Scientific Chemicals

2'-Chloro-5'-(trifluoromethyl)acetophenone, 96%, Thermo Scientific Chemicals

Catalog number: B23524.03
1 g, Each
Thermo Scientific Chemicals

2'-Chloro-5'-(trifluoromethyl)acetophenone, 96%, Thermo Scientific Chemicals

Catalog number: B23524.03
1 g, Each
Quantity
Catalog number: B23524.03
also known as B23524-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71648-45-8
IUPAC Name
1-[2-chloro-5-(trifluoromethyl)phenyl]ethan-1-one
Molecular Formula
C9H6ClF3O
InChI Key
YRGBMTWHOFQSDJ-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC(=CC=C1Cl)C(F)(F)F
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥95.0%
Identification (FTIR)
Conforms
Refractive Index
1.4755-1.4805 @ 20?C

Description

2'-Chloro-5'-(trifluoromethyl)acetophenone is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Chloro-5′-(trifluoromethyl)acetophenone is used as a pharmaceutical intermediate.

Solubility
Sparingly soluble in water 0.11g/L @ 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text