Thermo Scientific Chemicals

2(3)-tert-Butyl-4-methoxyphenol, 96%, Thermo Scientific Chemicals

Catalog number: B23530.18
50 g, Each
Thermo Scientific Chemicals

2(3)-tert-Butyl-4-methoxyphenol, 96%, Thermo Scientific Chemicals

Catalog number: B23530.18
50 g, Each
Quantity
Catalog number: B23530.18
also known as B23530-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25013-16-5
IUPAC Name
2-tert-butyl-4-methoxyphenol
Molecular Formula
C11H16O2
InChI Key
MRBKEAMVRSLQPH-UHFFFAOYSA-N
SMILES
COC1=CC=C(O)C(=C1)C(C)(C)C
Appearance (Color)
White to cream to pale yellow
Form
Crystals or powder or crystalline powder or flakes
Assay (GC)
≥95.0%

Description

2(3)-tert-Butyl-4-methoxyphenol is used as an industrial antioxidant. It is used as a food additive, preservative in food packaging, animal feed, cosmetics, rubber, and petroleum products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2(3)-tert-Butyl-4-methoxyphenol is used as an industrial antioxidant. It is used as a food additive, preservative in food packaging, animal feed, cosmetics, rubber, and petroleum products.

Solubility
Soluble in ethanol, methanol, chloroform, fats, and oils. Insoluble in water.

Notes
Store in the dark place. Store away from light, oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text