2-Ethylhexyl bromide is used in the synthesis of methoxy-4-(2-ethylhexyloxy)benzene, 1-bromo-4-(2?-ethylhexyloxy)benzene and 3,6-diaryl substituted 9-alkylcarbazoles as hole transporting materials for various organic light emitting devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Ethylhexyl bromide is used in the synthesis of methoxy-4-(2-ethylhexyloxy)benzene, 1-bromo-4-(2′-ethylhexyloxy)benzene and 3,6-diaryl substituted 9-alkylcarbazoles as hole transporting materials for various organic light emitting devices.
Solubility
Soluble in chloroform, ethyl acetate. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from heat, oxidizing agents.
RUO – Research Use Only
Neef CJ and Ferraris JP. MEH-PPV: improved synthetic procedure and molecular weight control. Macromolecules. 2000, 33 (7), 2311-14.
Lijun Huo; Ye Huang, Benhu Fan; Xia Guo; Yan Jing; Maojie Zhang; Yongfang Li; Jianhui Hou. Synthesis of a 4,8-dialkoxy-benzo[1,2-b:4,5-b’]difuran unit and its application in photovoltaic polymer. Chemical Communications. 2012, 48 (27), 3318-3320.