Thermo Scientific Chemicals

2,4,6-Trimethoxybenzonitrile, 98%, Thermo Scientific Chemicals

Catalog number: B23540.06
5 g, Each
Thermo Scientific Chemicals

2,4,6-Trimethoxybenzonitrile, 98%, Thermo Scientific Chemicals

Catalog number: B23540.06
5 g, Each
Quantity
Catalog number: B23540.06
also known as B23540-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2571-54-2
IUPAC Name
2,4,6-trimethoxybenzonitrile
Molecular Formula
C10H11NO3
InChI Key
GBRHJUMDNWLSCT-UHFFFAOYSA-N
SMILES
COC1=CC(OC)=C(C#N)C(OC)=C1
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
139.5-145.5?C
Assay (GC)
≥97.5%
Appearance (Color)
White to pale brown or pale orange

Description

2,4,6-Trimethoxybenzonitrile is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethoxybenzonitrile is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text