Thermo Scientific Chemicals

DL-Pipecolinic acid, 99%, Thermo Scientific Chemicals

Catalog number: B23541.22
100 g, Each
Thermo Scientific Chemicals

DL-Pipecolinic acid, 99%, Thermo Scientific Chemicals

Catalog number: B23541.22
100 g, Each
Quantity
Catalog number: B23541.22
also known as B23541-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
535-75-1
IUPAC Name
piperidine-2-carboxylic acid
Molecular Formula
C6H11NO2
InChI Key
HXEACLLIILLPRG-UHFFFAOYNA-N
SMILES
OC(=O)C1CCCCN1
Assay (Non-aqueous acid-base Titration)
≥98.5% to ≤101.5%
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale yellow or pale cream to pale brown
Identification (FTIR)
Conforms

Description

DL-Pipecolinic acid is used as a diagnostic marker of pyridoxine-dependent epilepsy. It acts as a ligand in copper-catalyzed approach for P-arylation of organophosphorus compounds. It is used as organocatalyst and novel substrate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Pipecolinic acid is used as a diagnostic marker of pyridoxine-dependent epilepsy. It acts as a ligand in copper-catalyzed approach for P-arylation of organophosphorus compounds. It is used as organocatalyst and novel substrate in organic synthesis.

Solubility
Soluble in water (50 mg/ml), and boiling alcohol.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text