Thermo Scientific Chemicals

3-Bromofuran, 97%, stab. with 0.5% calcium carbonate, Thermo Scientific Chemicals

Catalog number: B23543.06
5 g, Each
Thermo Scientific Chemicals

3-Bromofuran, 97%, stab. with 0.5% calcium carbonate, Thermo Scientific Chemicals

Catalog number: B23543.06
5 g, Each
Quantity
Catalog number: B23543.06
also known as B23543-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22037-28-1
Form
Liquid
Appearance (Color)
Clear colorless to brown
Assay (GC)
>96.0%
Refractive Index
1.4935-1.4985 @ 20?C
Comment
Stabilised with 0.5% Calcium carbonate

Description

3-Bromofuran was a synthesis reagent used in the preparation of 2-substituted 3-furfurals. It was also used in the synthesis of 5,6-dehydronorcantharidins. It is one of the volatile metabolites discussed in studies for detecting and discriminating diseases of onion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromofuran was a synthesis reagent used in the preparation of 2-substituted 3-furfurals. It was also used in the synthesis of 5,6-dehydronorcantharidins. It is one of the volatile metabolites discussed in studies for detecting and discriminating diseases of onion.

Solubility
Soluble in chloroform, ethyl acetate. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from heat, oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text