Thermo Scientific Chemicals

1,2-Dimethylimidazole, 98%, Thermo Scientific Chemicals

Catalog number: B23544.36
500 g, Each
Thermo Scientific Chemicals

1,2-Dimethylimidazole, 98%, Thermo Scientific Chemicals

Catalog number: B23544.36
500 g, Each
Quantity
Catalog number: B23544.36
also known as B23544-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1739-84-0
IUPAC Name
1,2-dimethyl-1H-imidazole
Molecular Formula
C5H8N2
InChI Key
GIWQSPITLQVMSG-UHFFFAOYSA-N
SMILES
CN1C=CN=C1C
Form
Crystals or powder or crystalline powder or fused solid or chunks
Assay (GC)
≥97.5%
Appearance (Color)
Colorless to white to pale yellow
Melting Point (clear melt)
27.0-40.0?C
Identification (FTIR)
Conforms

Description

1,2-Dimethylimidazole is used in the synthesis of 1,2-dimethyl-3-n-butylimidazoliumchloride and 1,2-dimethyl-3-n-propylimidazolium chloride. It also can be used in the synthesis of 1-(2-methoxyethyl)-2,3-dimethylimidazolium chloride and hexafluorophosphate salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dimethylimidazole is used in the synthesis of 1,2-dimethyl-3-n-butylimidazoliumchloride and 1,2-dimethyl-3-n-propylimidazolium chloride. It also can be used in the synthesis of 1-(2-methoxyethyl)-2,3-dimethylimidazolium chloride and hexafluorophosphate salts.

Solubility
Soluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text