Thermo Scientific Chemicals

3-Hydroxyphenylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: B23563.06
5 g, Each
Thermo Scientific Chemicals

3-Hydroxyphenylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: B23563.06
5 g, Each
Quantity
Catalog number: B23563.06
also known as B23563-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
621-37-4
IUPAC Name
2-(3-hydroxyphenyl)acetic acid
Molecular Formula
C8H8O3
InChI Key
FVMDYYGIDFPZAX-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=CC(O)=C1
Appearance (Color)
Pale cream to cream to pale brown to pale gray
Assay (Silylated GC)
≥98.5%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
127.0-133.0?C
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text