3-Hydroxyphenylacetic acid, 99%, Thermo Scientific Chemicals
3-Hydroxyphenylacetic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Hydroxyphenylacetic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number B23563.14
also known as B23563-14
Price (USD)/ Each
245.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
245.00
Add to cart
3-Hydroxyphenylacetic acid, 99%, Thermo Scientific Chemicals
Catalog numberB23563.14
Price (USD)/ Each
245.00
-
Add to cart
Chemical Identifiers
CAS621-37-4
IUPAC Name2-(3-hydroxyphenyl)acetic acid
Molecular FormulaC8H8O3
InChI KeyFVMDYYGIDFPZAX-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to pale brown to pale gray
Assay (Silylated GC)≥98.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)127.0-133.0?C
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only