Thermo Scientific Chemicals

3-Methoxybenzoyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: B23571.30
250 g, Each
Thermo Scientific Chemicals

3-Methoxybenzoyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: B23571.30
250 g, Each
Quantity
Catalog number: B23571.30
also known as B23571-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1711-05-3
IUPAC Name
3-methoxybenzoyl chloride
Molecular Formula
C8H7ClO2
InChI Key
RUQIUASLAXJZIE-UHFFFAOYSA-N
SMILES
COC1=CC=CC(=C1)C(Cl)=O
Refractive Index
1.5575-1.5605 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Form
Liquid

Description

3-Methoxybenzoyl chloride is used in the synthesis of 2-arylbenzofuran-based molecules that act against Alzheimer’s disease, combating symptoms. In addition, it aids in the optimization of dihydropyrrolopyrimidine inhibitors against PI3K (phosphoinositide 3-kinase) resulting in tumor growth inhibition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform. Reacts with water.

Notes
Moisture Sensitive. Store under dry inert gas. Store away from moisture, strong bases and oxidizing agents. Protect from humidity and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text