Thermo Scientific Chemicals

8-Nitroquinoline, 98%, Thermo Scientific Chemicals

Catalog number: B23574.22
100 g, Each
Thermo Scientific Chemicals

8-Nitroquinoline, 98%, Thermo Scientific Chemicals

Catalog number: B23574.22
100 g, Each
Quantity
Catalog number: B23574.22
also known as B23574-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
607-35-2
IUPAC Name
8-nitroquinoline
Molecular Formula
C9H6N2O2
InChI Key
OQHHSGRZCKGLCY-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=C2N=CC=CC2=CC=C1
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
86.0-93.0°C
Appearance (Color)
Pale yellow or pale yellowish-brown
Assay (GC)
≥97.5%

Description

8-Nitroquinoline is used in the preparation of bisubstituted phenoxy-6-methoxy-8-aminoquinoline. It is used to prepare furazano [3,4-h] quinoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Nitroquinoline is used in the preparation of bisubstituted phenoxy-6-methoxy-8-aminoquinoline. It is used to prepare furazano [3,4-h] quinoline.

Solubility
Soluble in ethanol, ethyl ether and benzene, slightly soluble in cold water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text