Thermo Scientific Chemicals

2-Chlorotoluene, 98%, Thermo Scientific Chemicals

Catalog number: B23596.22
100 g, Each
Thermo Scientific Chemicals

2-Chlorotoluene, 98%, Thermo Scientific Chemicals

Catalog number: B23596.22
100 g, Each
Quantity
Catalog number: B23596.22
also known as B23596-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95-49-8
IUPAC Name
1-chloro-2-methylbenzene
Molecular Formula
C7H7Cl
InChI Key
IBSQPLPBRSHTTG-UHFFFAOYSA-N
SMILES
CC1=CC=CC=C1Cl
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Refractive Index
1.5235-1.5265 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms

Description

2-Chlorotoluene is used for permanganate oxidation to give o-chlorobenzoic acid. It is used in the preparation of 3-chloro-4-methylcatechol and 4-chloro-3-methylcatechol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorotoluene is used for permanganate oxidation to give o-chlorobenzoic acid. It is used in the preparation of 3-chloro-4-methylcatechol and 4-chloro-3-methylcatechol.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text