Thermo Scientific Chemicals

O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98%, Thermo Scientific Chemicals

Catalog number: B23597.14
25 g, Each
Thermo Scientific Chemicals

O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98%, Thermo Scientific Chemicals

Catalog number: B23597.14
25 g, Each
Quantity
Catalog number: B23597.14
also known as B23597-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
94790-37-1
IUPAC Name
(1H-1,2,3-benzotriazol-1-yl)[bis(dimethylamino)methylidene]oxidanium
Molecular Formula
C11H16N5O
InChI Key
CLZISMQKJZCZDN-UHFFFAOYSA-N
SMILES
CN(C)C(=[O+]N1N=NC2=CC=CC=C12)N(C)C
Identification (FTIR)
Conforms
Form
Crystalline powder
Assay (HPLC)
≥97.5%
Loss on Drying
<1.0%
Appearance (Color)
White to pale cream

Description

O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate is used as a coupling reagent for solid-phase peptide synthesis. It has been shown to effectively suppress racemization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-(1H-Benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate is used as a coupling reagent for solid-phase peptide synthesis. It has been shown to effectively suppress racemization.

Solubility
Soluble in acetonitrile.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text