4-Dimethylamino-2-methoxybenzaldehyde is used to produce 2,6-Dichloro-N'-(4-dimethylamino-2-methoxy-benzylidene)-benzene-1,4-diamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Dimethylamino-2-methoxybenzaldehyde is used to produce 2,6-Dichloro-N′-(4-dimethylamino-2-methoxy-benzylidene)-benzene-1,4-diamine.
Solubility
Slightly soluble 0.32 g/L at 25°C.
Notes
Air sensitive. Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents.
RUO – Research Use Only
Jack W. Rasburn and Frederick H. C. Stewart. 431. The action of tin and hydrochloric acid on p-dialkylaminobenzaldehydes. Part II. The preparation of some new anthracenes. Journal of the Chemical Society. 1957, 2237-2239.