Thermo Scientific™

3-Cyclohexylpropionic acid, 98+%, Thermo Scientific Chemicals

Catalog number: B23613.18
50 g, Each
Thermo Scientific™

3-Cyclohexylpropionic acid, 98+%, Thermo Scientific Chemicals

Catalog number: B23613.18
50 g, Each
Quantity
Catalog number: B23613.18
also known as B23613-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
701-97-3
IUPAC Name
3-cyclohexylpropanoic acid
Molecular Formula
C9H16O2
InChI Key
HJZLEGIHUQOJBA-UHFFFAOYSA-N
SMILES
OC(=O)CCC1CCCCC1
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Refractive Index
1.4620-1.4660 @ 20?C
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Assay (Silylated GC)
≥98.0%

Description

3-Cyclohexylpropionic acid can be used to produce 3-cyclohexyl-propionyl chloride. It is used as pharmaceutical intermediates in organic synthesis. It can also be used for the synthesis of pineapple esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Cyclohexylpropionic acid can be used to produce 3-cyclohexyl-propionyl chloride. It is used as pharmaceutical intermediates in organic synthesis. It can also be used for the synthesis of pineapple esters.

Solubility
Soluble in water 10g/ L.

Notes
Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text