Copper(II) acetate, anhydrous, 98%, Thermo Scientific Chemicals
Copper(II) acetate, anhydrous, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper(II) acetate, anhydrous, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number B23615.22
also known as B23615-22
Price (USD)/ Each
169.65
Online exclusive
188.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
169.65
Online exclusive
188.00 
Save 18.35 (10%)
Add to cart
Copper(II) acetate, anhydrous, 98%, Thermo Scientific Chemicals
Catalog numberB23615.22
Price (USD)/ Each
169.65
Online exclusive
188.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS142-71-2
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>97.5%
Appearance (Color)Blue-green
Elemental AnalysisCl:<200ppm
pH5.0-5.3
Formpowder
View more
Copper(II) acetate is used in biochemical applications such as DNA extraction. It is used as a source of copper in inorganic synthesis, an oxidizing agent and as a catalyst in organic synthesis. It is also used to couple two terminal alkynes to make a 1,3-diyne. It is widely used in pigments, manufacture of paris green, in textile dyeing, skin conditioner in cosmetics, and as antioxidant and stabilizer in food grade polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) acetate is used in biochemical applications such as DNA extraction. It is used as a source of copper in inorganic synthesis, an oxidizing agent and as a catalyst in organic synthesis. It is also used to couple two terminal alkynes to make a 1,3-diyne. It is widely used in pigments, manufacture of paris green, in textile dyeing, skin conditioner in cosmetics, and as antioxidant and stabilizer in food grade polymers.

Solubility
Slightly soluble in water. Soluble in alcohol and ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Mansour, A. M. What happens when (1H-benzimidazol-2-ylmethyl)-N-phenyl amine is added to copper(II) acetate? Spectroscopic, magnetic, and DFT studies. Inorg. Chim. Acta. 2013, 408, 186-192.
  2. Abdalla, K.; Rahmat, A.; Azizan, A. Effect of copper (II) acetate pretreatment on zinc phosphate coating morphology and corrosion resistance. J. Coat. Technol. Res. 2013, 10 (1), 133-139.