Thermo Scientific Chemicals

N-Acetyl-L-glutamic acid, 99%

Catalog number: B23621.14
25 g, Each
Thermo Scientific Chemicals

N-Acetyl-L-glutamic acid, 99%

Catalog number: B23621.14
25 g, Each
Quantity
Catalog number: B23621.14
also known as B23621-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1188-37-0
IUPAC Name
2-acetamidopentanedioic acid
Molecular Formula
C7H11NO5
InChI Key
RFMMMVDNIPUKGG-UHFFFAOYNA-N
SMILES
CC(=O)NC(CCC(O)=O)C(O)=O
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Appearance (Color)
White
Optical Rotation
-13 ° to -18° (c=1 in water)
Form
Crystals or powder or crystalline powder

Description

N-Acetyl-L-glutamic acid is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-L-glutamic acid is used as pharmaceutical intermediates.

Solubility
Soluble in water. 2.7 g/100 mL (20°C)

Notes
Store at 4°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text