N-Acetyl-L-glutamic acid, 99%
N-Acetyl-L-glutamic acid, 99%
Thermo Scientific Chemicals

N-Acetyl-L-glutamic acid, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number B23621.22
also known as B23621-22
Price (USD)/ Each
67.80
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
67.80
Add to cart
N-Acetyl-L-glutamic acid, 99%
Catalog numberB23621.22
Price (USD)/ Each
67.80
-
Add to cart
Chemical Identifiers
CAS1188-37-0
IUPAC Name2-acetamidopentanedioic acid
Molecular FormulaC7H11NO5
InChI KeyRFMMMVDNIPUKGG-UHFFFAOYNA-N
SMILESCC(=O)NC(CCC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Appearance (Color)White
Optical Rotation-13 ° to -18° (c=1 in water)
FormCrystals or powder or crystalline powder
N-Acetyl-L-glutamic acid is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-L-glutamic acid is used as pharmaceutical intermediates.

Solubility
Soluble in water. 2.7 g/100 mL (20°C)

Notes
Store at 4°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only
Joseph V. Auditore; Littleton Wade and Erik J. Olson. Occurrence of N-Acetyl-L-Glutamic Acid in the Human Brain. Journal of Neurochemistry. 1966, 13, 1149-1155.