Thermo Scientific Chemicals

6-Acetamidohexanoic acid hydrate, 99%, Thermo Scientific Chemicals

Catalog number: B23623.14
25 g, Each
Thermo Scientific Chemicals

6-Acetamidohexanoic acid hydrate, 99%, Thermo Scientific Chemicals

Catalog number: B23623.14
25 g, Each
Quantity
Catalog number: B23623.14
also known as B23623-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57-08-9
IUPAC Name
6-acetamidohexanoic acid
Molecular Formula
C8H15NO3
InChI Key
WDSCBUNMANHPFH-UHFFFAOYSA-N
SMILES
CC(=O)NCCCCCC(O)=O
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%, dry wt. basis
Water Content (Karl Fischer Titration)
≤10.0%
Appearance (Color)
White
Form
Powder
Comment
MP recorded on dried material

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text