Thermo Scientific Chemicals

2-Nitrobenzenesulfenyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: B23627.18
50 g, Each
Thermo Scientific Chemicals

2-Nitrobenzenesulfenyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: B23627.18
50 g, Each
Quantity
Catalog number: B23627.18
also known as B23627-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7669-54-7
IUPAC Name
[(2-nitrophenyl)sulfanyl]chlorane
Molecular Formula
C6H4ClNO2S
InChI Key
NTNKNFHIAFDCSJ-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=CC=C1SCl
Appearance (Color)
Yellow
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
70.0-79.0?C

Description

2-Nitrobenzenesulfenyl chloride is used as a reagent for N-protection as N-2-nitrophenylsulfenyl (Nps) derivatives, in amino acids and peptides and nucleosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrobenzenesulfenyl chloride is used as a reagent for N-protection as N-2-nitrophenylsulfenyl (Nps) derivatives, in amino acids and peptides and nucleosides.

Solubility
Reacts with water.

Notes
Store under dry inert gas. Store away from moisture, strong bases and oxidizing agents. Protect from humidity and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text