Thermo Scientific Chemicals

5-Amino-2-methylbenzothiazole, 99%, Thermo Scientific Chemicals

Catalog number: B23631.09
10 g, Each
Thermo Scientific Chemicals

5-Amino-2-methylbenzothiazole, 99%, Thermo Scientific Chemicals

Catalog number: B23631.09
10 g, Each
Quantity
Catalog number: B23631.09
also known as B23631-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13382-43-9
IUPAC Name
2-methyl-1,3-benzothiazol-5-amine
Molecular Formula
C8H8N2S
InChI Key
GPWQHYMVUZYWIK-UHFFFAOYSA-N
SMILES
CC1=NC2=CC(N)=CC=C2S1
Form
Powder
Melting Point (clear melt)
97.0-103.0?C
Appearance (Color)
Pale orange to orange to brown
Assay (GC)
≥98.5%

Description

5-Amino-2-methylbenzothiazole is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Amino-2-methylbenzothiazole is used as pharmaceutical intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text