Thermo Scientific Chemicals

3-(Aminomethyl)pyridine, 98+%, Thermo Scientific Chemicals

Catalog number: B23634.22
100 g, Each
Thermo Scientific Chemicals

3-(Aminomethyl)pyridine, 98+%, Thermo Scientific Chemicals

Catalog number: B23634.22
100 g, Each
Quantity
Catalog number: B23634.22
also known as B23634-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3731-52-0
IUPAC Name
1-(pyridin-3-yl)methanamine
Molecular Formula
C6H8N2
InChI Key
HDOUGSFASVGDCS-UHFFFAOYSA-N
SMILES
NCC1=CC=CN=C1
Assay (GC)
≥98.0%
Refractive Index
1.5490-1.5530 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Identification (FTIR)
Conforms

Description

It finds its application as a rubefacient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a rubefacient.

Solubility
Fully miscible in water. Soluble in chloroform, ethyl acetate and methanol.

Notes
Air sensitive. Store away from air, carbon dioxide and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text